accessibility menu, dialog, popup

UserName

CAS RN 343-27-1

HClCH3ONHH3CN

CAS RN 343-27-1

IUPAC Name: hydrogen 7-methoxy-1-methyl-9H-pyrido[3,4-b]indole chloride
Synonyms: hydrogen harmine chloride
Molecular Weight (g/mol): 248.71
Molecular Formula: C13H13ClN2O
InChi Key: VNPLYCKZIUTKJM-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=CC2=C(C=C1)C1=C(N2)C(C)=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results