missing translation for 'onlineSavingsMsg'
Learn More

Harmine Hydrochloride 98.0+%, TCI America™

Catalog No. H0002100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0002100MG 100 mg
H00021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0002100MG Supplier TCI America Supplier No. H0002100MG
Only null left

Chemical Identifiers

CAS 343-27-1
Molecular Formula C13H13ClN2O
Molecular Weight (g/mol) 248.71
MDL Number MFCD00012641
InChI Key VNPLYCKZIUTKJM-UHFFFAOYSA-N
Synonym Banisterine Hydrochloride
PubChem CID 5359389
IUPAC Name hydrogen 7-methoxy-1-methyl-9H-pyrido[3,4-b]indole chloride
SMILES [H+].[Cl-].COC1=CC2=C(C=C1)C1=C(N2)C(C)=NC=C1

Specifications

Color White-Yellow
Quantity 100 mg
UN Number 1544
Formula Weight 248.71
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Harmine Hydrochloride
RTECSNumber MG9450000
TSCA No