Learn More
Hardy Diagnostics™ Standard Methods Agar
$445.12 - $459.47
Specifications
| Media Type | Plate count agar/Tryptone glucose yeast (TGY) agar |
|---|---|
| For Use With (Application) | Cultivation and enumeration of microorganisms in dairy products, food, and water |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
02-686-213
|
Hardy Diagnostics™
U295 |
12/Pk. |
Pack of 12 Each for $445.12
|
|
|||||
|
02-686-214
|
Hardy Diagnostics™
U95 |
20/Pk. |
Pack of 20 Each for $459.47
|
|
|||||
Specifications
| Plate count agar/Tryptone glucose yeast (TGY) agar | |
| Cultivation and enumeration of microorganisms in dairy products, food, and water |