Learn More
Hardy Diagnostics™ Standard Methods Agar
Supplier:  Hardy Diagnostics™ U95
 View more versions of this product
Catalog No. 02-686-214
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Plate count agar/Tryptone glucose yeast (TGY) agar
Cultivation and enumeration of microorganisms in dairy products, food, and water
20/Pk.
Product Suggestions
Product Suggestions