missing translation for 'onlineSavingsMsg'
Learn More
Learn More
- Home
- Products
- Facility Safety and Maintenance
- Labels and Label Makers
- Custom Labels
- Brady™ Hard Hat Labels - Pressure Sensitive Vinyl
Brady™ Hard Hat Labels - Pressure Sensitive Vinyl
Catalog No. 19110287
Change view
Click to view available options
Color:
Black on White
Black on Yellow
Black/Blue/Red on White
Black/Blue/Red/Yellow on White
Black/Brown on White
Black/Orange on White
Black/Red on White
Black/Red on Yellow
Black/Yellow on White
Blue on White
Green on White
Red on White
White on Green
White on Red
Display Text:
ACCIDENT FREE 5 YEARS AWARD
ACCIDENT FREE AWARD 1 YEAR AWARD
ACCIDENT PREVENTION MEMBER
AUTHORIZED FORKLIFT DRIVER
AUTHORIZED OPERATOR FORK LIFT
BASIC LIFE SUPPORT CERTIFIED CPR
BE CAREFUL
CERTIFIED CRANE OPERATOR
CERTIFIED FIRST AID
CERTIFIED FORKLIFT DRIVER
CHEMICAL EMERGENCY TEAM
CPR CERTIFIED
CPR FIRST AID CERTIFIED AND TRAINED
CPR TRAINED CERTIFIED
EMERGENCY MEDICAL TECHNICIAN
EMERGENCY RESPONSE TEAM
ERGONOMICS COMMITTEE
FIRST AID SQUAD
FIRST... SAFETY ...LAST
FORKLIFT SAFETY TRAINED
GOOD HOUSEKEEPING COMMITTEE
HAZ MAT SQUAD
HAZARDOUS MATERIALS MANAGEMENT
HEALTH & SAFETY COMMITTEE MEMBER
HEALTH & SAFETY COMMITTEE
HEALTH AND SAFETY/COMMITTEE MEMBER
KNOCK OUT... ACCIDENTS
LOCK-OUT TAG-OUT TRAINED
MEDICAL EMERGENCY TEAM
MEMBER FIRE BRIGADE
OUR AIM... NO ACCIDENTS
PLANT SAFETY MEMBER
QUALIFIED CRANE OPERATOR
QUALIFIED FORK LIFT OPERATOR
QUALITY STARTS WITH ME!
RESCUE TEAM
RIGHT TO KNOW TRAINED
SAFE DRIVER
SAFE WORKER NO ACCIDENTS
SAFE WORKER NO ACCIDENTS 5 YEARS
SAFE WORKER NO ACCIDENTS/1 YEAR
SAFETY AWARD WINNER
SAFETY BEGINS WITH TEAMWORK
SAFETY IS NOT AN ACCIDENT
SAFETY NEVER HURTS
SAFETY OBSERVER
SAFETY REPRESENTATIVE
SAFETY SUPERVISOR
SAFETY... IT'S MY JOB
THINK SAFETY 1st
VISITOR
YOUR FUTURE BRIGHTENS WITH SAFETY
54 product options available for selection
| Catalog No. | Color | Display Text |
|---|---|---|
| 19110287 | Black/Red on Yellow | SAFETY NEVER HURTS |
| 19110268 | White on Green | BE CAREFUL |
| 19110265 | Black on Yellow | AUTHORIZED OPERATOR FORK LIFT |
| 19110266 | Black on Yellow | AUTHORIZED FORKLIFT DRIVER |
| 19110270 | Black on Yellow | CERTIFIED FORKLIFT DRIVER |
| 19110271 | White on Red | CPR FIRST AID CERTIFIED AND TRAINED |
| 19110272 | Black/Red on White | EMERGENCY RESPONSE TEAM |
| 19110274 | Black/Red on White | FIRST... SAFETY ...LAST |
| 19110276 | Green on White | HEALTH & SAFETY COMMITTEE |
| 19110275 | Black on Yellow | FORKLIFT SAFETY TRAINED |
| 19110277 | Green on White | HEALTH & SAFETY COMMITTEE MEMBER |
| 19110278 | Black/Brown on White | KNOCK OUT... ACCIDENTS |
| 19110279 | Black/Red on White | LOCK-OUT TAG-OUT TRAINED |
| 19110280 | Black/Red on White | OUR AIM... NO ACCIDENTS |
| 19110281 | Red on White | QUALITY STARTS WITH ME! |
| 19110282 | Black/Orange on White | QUALIFIED CRANE OPERATOR |
| 19110285 | Black/Red on White | SAFETY... IT'S MY JOB |
| 19110283 | Black/Yellow on White | QUALIFIED FORK LIFT OPERATOR |
| 19110284 | Black/Yellow on White | SAFETY BEGINS WITH TEAMWORK |
| 19110286 | Black/Red on Yellow | SAFETY IS NOT AN ACCIDENT |
| 19110288 | Red on White | THINK SAFETY 1st |
| 19110289 | Black on White | VISITOR |
| 19110290 | Black/Yellow on White | YOUR FUTURE BRIGHTENS WITH SAFETY |
| 19110291 | Black/Red on White | CERTIFIED CRANE OPERATOR |
| 19110292 | Black/Blue/Red on White | CPR TRAINED CERTIFIED |
| 19110293 | Black/Blue/Red/Yellow on White | RIGHT TO KNOW TRAINED |
| 19110295 | Green on White | ACCIDENT FREE AWARD 1 YEAR AWARD |
| 19110296 | Green on White | ACCIDENT FREE 5 YEARS AWARD |
| 19110297 | Green on White | SAFE WORKER NO ACCIDENTS/1 YEAR |
| 19110298 | Green on White | SAFE WORKER NO ACCIDENTS 5 YEARS |
| 19110299 | Green on White | SAFETY AWARD WINNER |
| 19110300 | Red on White | EMERGENCY RESPONSE TEAM |
| 19110301 | Green on White | SAFE DRIVER |
| 19110303 | Red on White | MEDICAL EMERGENCY TEAM |
| 19110302 | Red on White | HAZARDOUS MATERIALS MANAGEMENT |
| 19110304 | Black on White | CERTIFIED CRANE OPERATOR |
| 19110305 | Red on White | CPR CERTIFIED |
| 19110306 | Red on White | CHEMICAL EMERGENCY TEAM |
| 19110308 | Green on White | SAFETY SUPERVISOR |
| 19110307 | Blue on White | GOOD HOUSEKEEPING COMMITTEE |
| 19110310 | Red on White | RESCUE TEAM |
| 19110309 | Red on White | CERTIFIED FIRST AID |
| 19110314 | Black on White | EMERGENCY MEDICAL TECHNICIAN |
| 19110313 | Green on White | HEALTH AND SAFETY/COMMITTEE MEMBER |
| 19110315 | Green on White | PLANT SAFETY MEMBER |
| 19110316 | Green on White | SAFETY OBSERVER |
| 19110317 | Green on White | SAFETY REPRESENTATIVE |
| 19110318 | Red on White | ACCIDENT PREVENTION MEMBER |
| 19110319 | Red on White | BASIC LIFE SUPPORT CERTIFIED CPR |
| 19110320 | Green on White | SAFE WORKER NO ACCIDENTS |
| 19110321 | Red on White | HAZ MAT SQUAD |
| 19110322 | Green on White | ERGONOMICS COMMITTEE |
| 19110326 | Red on White | MEMBER FIRE BRIGADE |
| 19110327 | Green on White | FIRST AID SQUAD |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 54 options available.
Description
- High gloss, durable, flexible vinyl with a permanent cold temperature pressure sensitive acrylic adhesive
- Good chemical, oil and water resistance
- Average outdoor durability 8-10 years
- Service temperature of -40°F to 180°F (-40°C to 82°C)
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Description | Label |
| Adhesive Type | Permanent Acrylic, Self-adhesive |
| Color | Black/Red on Yellow |
| Format | Label |
| Material | Vinyl |
| Printable Sides | Single-side Printable |
| Product Type | Label |
| Display Text | SAFETY NEVER HURTS |
| Languages | English |
| No. per Pack | 4 Labels/Card |
| Quantity | 1 Card |
| Shape | Round |
| Width (Metric) | 69.85 mm |
| Diameter (English) | 2 in. |
| Show More Show Less |