Learn More
Brady™ Hard Hat Labels - Pressure Sensitive Vinyl
Pressure Sensitive Vinyl (B-946) is an indoor/outdoor marking film ideal for pipe marking, arrow tape, vehicle placards, pedestal markers, warning panels and safety signs.
Supplier:  Brady™ 42243
 View more versions of this product
Catalog No. 19110275
Only null left
Description
Description
  • High gloss, durable, flexible vinyl with a permanent cold temperature pressure sensitive acrylic adhesive
  • Good chemical, oil and water resistance
  • Average outdoor durability 8-10 years
  • Service temperature of -40°F to 180°F (-40°C to 82°C)
Specifications
Chemical Identifiers
Not Autoclavable
Label
Single-side Printable
English
1 Card
Label
Vinyl
Label
4 Labels/Card
2 in.
Specifications
Not Autoclavable
Permanent Acrylic, Self-adhesive
Label
Single-side Printable
FORKLIFT SAFETY TRAINED
4 Labels/Card
Rectangle
2 in.
Label
Black on Yellow
Vinyl
Label
English
1 Card
69.85 mm
Product Suggestions
Product Suggestions