missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Hapten 4-hydroxy-3-nitrophenyl acetyl NP Chimeric Recombinant Mouse Monoclonal Antibody (B1-8)
GREENER_CHOICE

Catalog No. MA551697
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551697 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551697 Supplier Invitrogen™ Supplier No. MA551697
Only null left

Mouse Recombinant Monoclonal Antibody

Hapten (4-hydroxy-3-nitropheny1) acetyl (NP).

Hapten 4-hydroxy-3-nitrophenyl acetyl NP is a small molecule that consists of a hapten, which is the 4-hydroxy-3-nitrophenyl acetyl group, linked to a carrier protein, which is NP or nitrophenol. The carrier protein is an important component of this molecule as it helps to elicit an immune response by presenting the hapten portion of the molecule to the immune system. NP is commonly used as a carrier in immunology research because it is a small, soluble molecule that is easily conjugated with haptens. When the hapten binds to specific antibodies, the carrier protein helps to stimulate an immune response and activate B cells, which can lead to the production of specific antibodies against the hapten.

Specifications

Antigen Hapten 4-hydroxy-3-nitrophenyl acetyl NP Chimeric
Applications Functional Assay
Classification Recombinant Monoclonal
Clone B1-8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Host Species Mouse
Immunogen 4-hydroxy-3-nitrophenyl acetyl (NP) - conjugated to NP12-chicken gammaglobulin (CG).
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b λ
Show More Show Less