missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate, Crystal, Reagent, ACS, 97.5%, Spectrum™ Chemical

Catalog No. 18600689
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600689 50 kg Amber Glass Bottle
18600682 25 g Amber Glass Bottle
18600683 125 g Amber Glass Bottle
18600684 500 g Amber Glass Bottle
18600686 2.5 kg Amber Glass Bottle
18600685 6 x 500 g Amber Glass Bottle
18600687 4 x 2.5 kg Amber Glass Bottle
18600688 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18600689 Supplier Spectrum Chemical Mfg Cor Supplier No. A127050KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A1270, 1762-95-4, NH4SCN

Spectrum™ Chemical Ammonium Thiocyanate, Crystal, Reagent, ACS is an inorganic laboratory reagent commonly used by the soft drinks industry to determine the iron content in soft drinks by colorimetry. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.12
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
IUPAC Name ammonium cyanosulfanide
SMILES [NH4+].[S-]C#N

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 50 kg
Percent Purity 97.5%
Grade ACS
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Ammonium Thiocyanate, Crystal