Learn More
Hanna™ Instruments pH Buffer Solutions
Catalog No. 1160370
Encompass
Change view
Click to view available options
Quantity:
230 mL
Catalog No. Quantity
1160370 230 mL
1 options
Catalog No. 1160370 Supplier Hanna Instruments CDA Inc Supplier No. HI 7004M
Only null left
Used for calibration
  • Available in different pH ranges and quantity
  • Calibrated according to NIST standards
  • With/without certificates of analysis
Specifications
Quantity 230 mL
Recommended Storage Keep container closed and protected from direct sunlight. Store at room temperature (+15° to +25°C).