missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Hank's Balanced Salt Solutions

Catalog No. MT21023CV
Change view
Click to view available options
Quantity:
6 x 500 mL
6 x 1 L
Concentration:
1 X
With Additives:
Calcium, Magnesium
Without Additives:
No Calcium, No Magnesium
No Calcium, No Magnesium, No Phenol Red
No Phenol Red
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration With Additives Without Additives
MT21023CV 6 x 500 mL 1 X Calcium, Magnesium No Phenol Red
MT21023CM 6 x 1 L 1 X Calcium, Magnesium No Phenol Red
MT21021CV 6 x 500 mL 1 X - No Calcium, No Magnesium
MT21020CV 6 x 500 mL 1 X Calcium, Magnesium -
MT21021CM 6 x 1 L 1 X - No Calcium, No Magnesium
MT21022CV 6 x 500 mL 1 X - No Calcium, No Magnesium, No Phenol Red
MT21022CM 6 x 1 L 1 X - No Calcium, No Magnesium, No Phenol Red
MT21020CM 6 x 1 L 1 X Calcium, Magnesium -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MT21023CV Supplier Corning™ Supplier No. 21023CV

Salt solutions are shipped at ambient temperature unless otherwise indicated

pH of 10X solutions must be adjusted following dilution to 1X concentration.

Specifications

Content And Storage 2° to 30°C
Concentration 1 X
Form Liquid
Product Type Hanks' Balanced Salt Solution
With Additives Calcium, Magnesium
Without Additives No Phenol Red
Formulation Calcium Chloride (anhydrous) 0.14g/L, Potassium Chloride 0.4g/L , Monopotassium Phosphate 0.06g/L, Magnesium Sulfate (anhydrous) 0.977g/L, Sodium Chloride 8g/L , Disodium Phosphate (anhydrous) 0.0477g/L, Sodium Bicarbonate 0.35g/L, D-Glucose 1g/L
pH 7.1 to 7.4
Quantity 6 x 500 mL