missing translation for 'onlineSavingsMsg'
Learn More

Hughes Handheld Eye, Face and Body Shower, Wall Mount

Catalog No. 18666653
Change view
Click to view available options
Mounting:
Wall
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
18666653 Wall
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18666653 Supplier Justrite International Supplier No. OPT500

Hughes Handheld Eye, Face and Body Shower features two 45-degree angled double nozzles for hazardous environments where there is a risk of small spillages of chemicals or dangerous substances.

Features:

  • Fitted with pop-off dust covers, a non-return valve and flow regulator
  • Complete with 4.9-feet (1.5-m) of reinforced hose, particularly useful for treating disabled personnel
  • Lock-on actuating button on squeeze grip activation for ease of use
  • Meets ANSI and EN 15154 water flow standards for an eye/face wash. Includes an inspection record tag for recording test and service history.

Compliance

Not ANSI Compliant

Warranty

1 Year

Specifications

Activation Squeeze Grip
Color White, Green, Red
Depth (English) 9 in.
Depth (Metric) 229 mm
Flow Rate 3 gpm
For Use With (Application) Emergency Shower and Eye Wash with Water
Height (English) 7 in.
Height (Metric) 178 mm
Includes 1 Unit
Inlet Connection (Metric) 0.5 in. (F) NPT- Hose
Material Polyarylamide Reinforced with Glass Fibers (Body), Powder Coated Brass (Nozzles), EPDM (Eye Cups)
Mounting Wall
Pressure 22 to 87 psi
Product Type Emergency Eye Wash
Width (English) 19 in.
Width (Metric) 475 mm
Show More Show Less