Learn More
Laxco™ Handheld Analog Clinical Refractometer

When you need reliable, accurate readings of your Urine samples, the Laxco™ Analog Industrial Clinical Refractometers rugged design is built to last. With the ability to read Serum Protein, Specific Gravity and Refractive Index all on a single scale, it makes it the ideal choice.
$337.39
Specifications
| Type | Clinical Refractometer |
|---|---|
| Scale Range | 1.000 to 1.050 Urine Specific Gravity, 1 to 12 g/dl Serum Protein |
| Refractive Index | 1.331 to 1.36 |
| Sample Volume | 1 mL |
| Calibration | Distilled Water |
Description
Refractometers have been used for measuring the concentration of solutions for more than a century. Their primary purpose is to measure the degree at which light bends as it passes through a medium, and is used in a wide range of applications and industries as a quality control and diagnostic tool.
Laxco Analog Refractometers are designed for simple operation and built with all a metal construction for durability. Its industrial design is the most commonly used design among refractometer manufactures as it is a cost-effective model to build.
Each unit is inspected in the United States by a factory qualified technician under a rigid quality control process to ensure its accuracy, and performance.
Key Features:
- Quickly calibrate the instrument with a turn of a screw, and distilled water
- Comfortable eyecup blocks out ambient light and allows for crystal clear viewing of readings
- Automatic temperature compensation prevents the need for constant re-calibration of the instrument. As the ambient temperature changes, the scale will automatically compensate for the difference in the degree of refraction. This ensures an accurate reading every time
- All-metal construction creates durability. NOTE: Model 12-561-340 comes without ATC
- Footrest allows the instrument to be set on a flat surface without the worry of it rolling off working surfaces
- Rubber hand grip is comfortable, easy to grasp. Its non-slip pattern grip reduces that chance of accidental dropping the instrument
- Double hinged daylight plates create two points of movement to allow for greater flexibility in movement; making it easy to add samples and clean
- Portable, easy to use design.
Clinical Models:
Quickly and accurately get an indication of basic health parameters such as dehydration and blood plasma protein levels. Laxco Analog Clinical Refractometers uses three-scales to measure Total Serum Protein, Urine Specific Gravity, and Refractive Index. They are ideal for use in quality control, research, and clinical laboratories.
- Urine Specific Gravity: 1.000ppt to 1.050ppt with granularity to measure down to 0.001ppt at a 0.002ppt degree of accuracy
- Serum Protein: 1g to 12g with granularity to measure down to 0.2g/100mL at a 0.2g/100mL degree of accuracy
- Refractive Index: 1.334nD to 1.360nD with granularity to measure down to .0005nD at a 0.0005nD degree of accuracy.
Warranty
5 Years
Specifications
| Clinical Refractometer | |
| 1.331 to 1.36 | |
| Distilled Water | |
| 0.0005 | |
| Linear | |
| Clinical | |
| IP54 | |
| 0.881 lb. | |
| 172 x 39 x 39 mm | |
| 39 mm | |
| 204 mm | |
| 39 mm |
| 1.000 to 1.050 Urine Specific Gravity, 1 to 12 g/dl Serum Protein | |
| 1 mL | |
| 0.002; 0.2 g/dl; 0.0005 | |
| 0.002; 0.2 g/dl; 0.0005 | |
| Ambient | |
| Calibration Screw Driver, Hard Sided Case, Applicator, Calibration Fluid, Manual | |
| 1 | |
| 0.4 kg | |
| 1.535 in. | |
| 6.772 in. | |
| 1.535 in. | |
| 5 years optical and mechanical, 1 year electrical from date of purchase |
Warranty: Does not apply to daylight plate assembly