missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAMP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149854
Description
HAMP Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
HAMP is involved in the maintenance of iron homeostasis, and it is necessary for the regulation of iron storage in macrophages, and for intestinal iron absorption. The preproprotein is post-translationally cleaved into mature peptides of 20, 22 and 25 amino acids, and these active peptides are rich in cysteines, which form intramolecular bonds that stabilize their beta-sheet structures. These peptides exhibit antimicrobial activity.Specifications
| HAMP | |
| Polyclonal | |
| Unconjugated | |
| HAMP | |
| HAMP; Hamp1; HEPC; Hepc1; Hepc20; Hepc25; Hepcidin; hepcidin antimicrobial peptide; hepcidin antimicrobial peptide 1; Hepcidin-20; Hepcidin-25; HFE2B; LEAP1; LEAP-1; liver-expressed antimicrobial peptide 1; PLTR; Putative liver tumor regressor; UNQ487/PRO1003 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 57817, 84604 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P81172, Q99MH3 | |
| HAMP | |
| Recombinant Hepcidin (Hepc) (Gly23-Thr84). | |
| 200 μL | |
| Primary | |
| Human, Rat | |
| Antibody |