Learn More
MilliporeSigma™ Supelco™ Hamilton™ Needles for RN Syringes, Cone Tip, Stainless Steel
Hamilton™ needles for RN syringes are bevel tip needles, designed for optimum septum penetration and to prevent septum coring.
Supplier:  MilliporeSigma™ Supelco™ 19151U
 View more versions of this product
Catalog No. 111010318
Only null left
Description
Description
Specifications
Specifications
Stainless Steel (Tip)
2 in.
6/Pk.
51 mm
Hamilton RN Syringes
22s G
Cone Tip
250 μL to 10 mL
5
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Hamilton is a registered trademark of Hamilton Co.