missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Hamilton™ Needles for LT and TLL Syringes, Metal Hub (N)

Catalog No. 111011673
Change view
Click to view available options
Needle Gauge:
22 G
28 G
Needle Point Style:
2
3
5
Product Type:
Bevel Tip
Blunt Tip
Cone Tip
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Needle Point Style Product Type
111011673 22 G 2 Bevel Tip
111011672 22 G 3 Blunt Tip
111010467 22 G 5 Cone Tip
111011596 28 G 2 Bevel Tip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111011673 Supplier MilliporeSigma™ Supelco™ Supplier No. 21746

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Material Stainless Steel (Tip)
Needle Gauge 22 G
Includes Metal Hub (N)
Length (English) Needle 2 in.
Product Type Bevel Tip
Quantity 6/Pk.
Length (Metric) Needle 51 mm
Needle Point Style 2
For Use With (Equipment) Hamilton LT and TLL Syringes

Hamilton is a registered trademark of Hamilton Co.