Learn More
Hamilton™ Kel-F Hub Sharp Non-Coring Needles (Luer Lock)
Kel-F hub (KF) needles can be used with Luer Tip (LT) and PTFE Luer Lock (TLL) syringes and connectors. Standard needle length is 2 in. (51 mm). Custom lengths and point styles are available.
$121.64 - $206.62
Specifications
| Autoclavable | Not Autoclavable |
|---|---|
| Material | Stainless Steel |
| Length (English) Needle | 2 in. |
| Product Type | Kel-F Hub Needle |
| For Use With (Equipment) | Luer-tip and PTFE Luer-lock Syringes and Connectors |
| Catalog Number | Mfr. No. | Needle Gauge | Needle Point Style | Diameter (English) Inner | Diameter (English) Outer | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Needle Gauge | Needle Point Style | Diameter (English) Inner | Diameter (English) Outer | Price | Quantity | ||
|
14-815-440
|
Hamilton™
HAM90112 |
12 G | Sharp, Bevel, Curved | 0.085 in. | 0.08 in. |
Pack of 6 Each for $126.87
|
|
||
|
14-815-445
|
Hamilton™
HAM90117 |
17 G | Sharp, Bevel, Curved | 0.042 in. | 0.057 in. |
Pack of 6 Each for $121.64
|
|
||
|
14-815-444
|
Hamilton™
HAM90116 |
16 G | Sharp, Bevel, Curved | 0.046 in. | 0.064 in. |
Pack of 6 Each for $155.53
|
|
||
|
14-815-438
|
Hamilton™
HAM90110 |
10 G | Sharp, Bevel, Curved | 0.105 in. | 0.1 in. |
Pack of 6 Each for $126.87
|
|
||
|
14-815-447
|
Hamilton™
HAM90119 |
19 G | Sharp, Bevel, Curved | 0.027 in. | 0.042 in. |
Pack of 6 Each for $121.64
|
|
||
|
14-815-448
|
Hamilton™
HAM90120 |
20 G | Sharp, Bevel, Curved | 0.02 in. | 0.035 in. |
Pack of 6 Each for $155.53
|
|
||
|
14-815-449
|
Hamilton™
HAM90121 |
21 G | Sharp, Bevel, Curved | 0.02 in. | 0.032 in. |
Pack of 6 Each for $155.53
|
|
||
|
14-815-446
|
Hamilton™
HAM90118 |
18 G | Sharp, Bevel, Curved | 0.033 in. | 0.05 in. |
Pack of 6 Each for $121.64
|
|
||
|
14-815-451
|
Hamilton™
HAM90123 |
23 G | Sharp, Bevel, Curved | 0.013 in. | 0.025 in. |
Pack of 6 Each for $155.53
|
|
||
|
14-815-453
|
Hamilton™
HAM90125 |
25 G | Sharp, Bevel, Curved | 0.01 in. | 0.01 in. |
Pack of 6 Each for $126.60
|
|
||
|
14-815-454
|
Hamilton™
HAM90126 |
26 G | Sharp, Bevel, Curved | 0.01 in. | 0.018 in. |
Pack of 6 Each for $171.15
|
|
||
|
14-815-455
|
Hamilton™
HAM90127 |
27 G | Sharp, Bevel, Curved | 0.008 in. | 0.016 in. |
Pack of 6 Each for $161.62
|
|
||
|
14-815-456
|
Hamilton™
HAM90128 |
28 G | Sharp, Bevel, Curved | 0.007 in. | 0.014 in. |
Pack of 6 Each for $206.62
|
|
||
|
14-815-458
|
Hamilton™
HAM90130 |
30 G | Sharp, Bevel, Curved | 0.006 in. | 0.012 in. |
Pack of 6 Each for $206.62
|
|
||
|
14-815-459
|
Hamilton™
HAM90131 |
31 G | Sharp, Bevel, Curved | 0.005 in. | 0.01 in. |
Pack of 6 Each for $161.62
|
|
||
|
14-815-461
|
Hamilton™
HAM90139 |
26s G | Tapered | 0.005 in. | 0.018 in. |
Pack of 6 Each for $126.87
|
|
||
|
14-824-12
|
Hamilton™
HAM90138 |
22s G | Tapered | 0.005 in. | 0.028 in. |
Pack of 6 Each for $199.96
|
|
||
|
14-824-13
|
Hamilton™
HAM90122 |
22 G | Sharp, Bevel, Curved | 0.016 in. | 0.028 in. |
Pack of 6 Each for $148.77
|
|
||
|
14-815-442
|
Hamilton™
HAM90114 |
14 G | Sharp, Bevel, Curved | 0.062 in. | 0.062 in. |
Pack of 6 Each for $128.06
|
|
||
Description
Kel-F Hub Compatible Syringe Terminations
- Luer Tip (LT)- The needles are removable and fit over a ground glass hub which is tapered in the shape of a male luer. The LT termination will accept most hypodermic needles but was designed specifically for use with Hamilton Kel-F needles. This termination increases the dead volume in the syringe, which may not be appropriate for some applications. Autoclavable when disassembled.
- PTFE Luer Lock (TLL)- This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations. The version with slots is used during the attachment to some instruments but does not effect the connection to needles. Autoclavable when disassembled, except on 25 mL and greater syringes. Repeated autoclaving will shorten syringe life.
- Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 31-10
Six needles per pack.
Specifications
| Not Autoclavable | |
| 2 in. | |
| Luer-tip and PTFE Luer-lock Syringes and Connectors |
| Stainless Steel | |
| Kel-F Hub Needle |