missing translation for 'onlineSavingsMsg'
Learn More

Haloperidol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017784
Change view
Click to view available options
Quantity:
400 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017784 400 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017784 Supplier Merck Emd Millipore Supplier No. PHR1724400MG
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Haloperidol is a major tranquilizer that is indicated for the treatment of schizophrenia, mania and neurological disorders including hyperkinesia. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 52-86-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C21H23 ClFNO2
MDL Number MFCD00051423
Synonym Haloperidol; 4-[4-(4-Chlorophenyl)-4-hydroxy-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone; 4-[4-(4-Chlorophenyl)-4-hydroxypiperidino]-4′-fluorobutyrophenone; 4-[4-(p-Chlorophenyl)-4-hydroxypiperidino]-4′-fluorobutyrophenone
Type Certified Reference Material
Quantity 400 mg
Formula Weight 375.86
Grade Certified Reference Material