missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Half-Finger Medical Nylon Glove Liners

Half-Finger Medical Nylon Glove Liners are constructed of high-quality, 100% continuous filament nylon and packaged in a secure box. Wear under a disposable glove for additional comfort, or to reduce hand fatigue from perspiration.

$71.97 - $71.97

Specifications

Applications Healthcare
Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
View More Specs

Products 1
Catalog Number Mfr. No. Gender Price Quantity  
Catalog Number Mfr. No. Gender Price Quantity  
19044156
View Documents
Wells Lamont™
M117W
Women's
Pack of 25 Each for $71.97
Case of 4 Pack for $287.88
Only null left
 
Description

Description

  • 100% continuous nylon filament creates an ideal barrier between hands and disposable gloves
  • Reduces allergies and adverse reactions that can be caused by prolonged exposure to latex
  • Packaged in a secure box so that liners remain clean
  • Fully launderable up to 20 times without shrinking
  • Half finger, regular cuff design
  • Non-sterile
  • Made in US
Specifications

Specifications

Healthcare
White
Ambidextrous
Nylon
Non-sterile
Reducing Latex Allerigies and Adverse Reactions
Box
Uncoated
Reusable
Yes
Medical Glove Liner
Half
Knit
M117
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'