missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Halcinonide

Catalog No. AC464980010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464980010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC464980010 Supplier Thermo Scientific Chemicals Supplier No. 464980010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3093-35-4
Molecular Formula C24H32ClFO5
Molecular Weight (g/mol) 454.96
MDL Number MFCD00866006
InChI Key MUQNGPZZQDCDFT-JNQJZLCISA-N
IUPAC Name (1S,2S,4R,8S,9S,11S,12R,13S)-8-(2-chloroacetyl)-12-fluoro-11-hydroxy-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icos-17-en-16-one
SMILES CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)CC[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CCl

Specifications

Packaging glass bottle
Certificate of Analysis Available
Shipping Condition normal conditions
Physical Form Powder
Color White to Yellow
Melting Point 264°C to 265°C
Solubility Soluble in acetone, chloroform and DMSO; Slightly soluble in benzene, ethanol, methanol and ethyl ether; Insoluble in hexanes and water
Type Reagent
Quantity 1 g
Formula Weight 454.96
Chemical Name or Material Halcinonide
Show More Show Less
Hazard Category GHS08: Health hazard
ShelfLife 3 years
storageNote1 Freezer -20°C
Recommended Storage Freezer -20°C

RUO – Research Use Only