missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hafnocene Dichloride 98.0+%, TCI America™
$90.48 - $1413.08
Chemical Identifiers
| CAS | 12116-66-4 |
|---|---|
| Molecular Formula | C10H10Cl2Hf |
| Molecular Weight (g/mol) | 379.58 |
| MDL Number | MFCD00001438,MFCD00001438 |
| InChI Key | CSEGCHWAMVIXSA-UHFFFAOYSA-L |
| Synonym | bis cyclopentadienyl hafniumdichloride |
| PubChem CID | 131674741 |
| IUPAC Name | hafnium(4+) bis(cyclopenta-2,4-dien-1-ide) dichloride |
| SMILES | [Cl-].[Cl-].[Hf+4].[CH-]1C=CC=C1.[CH-]1C=CC=C1 |
Chemical Identifiers
| 12116-66-4 | |
| 379.58 | |
| CSEGCHWAMVIXSA-UHFFFAOYSA-L | |
| 131674741 | |
| [Cl-].[Cl-].[Hf+4].[CH-]1C=CC=C1.[CH-]1C=CC=C1 |
| C10H10Cl2Hf | |
| MFCD00001438,MFCD00001438 | |
| bis cyclopentadienyl hafniumdichloride | |
| hafnium(4+) bis(cyclopenta-2,4-dien-1-ide) dichloride |
Specifications
| 12116-66-4 | |
| C10H10Cl2Hf | |
| 1 g | |
| CSEGCHWAMVIXSA-UHFFFAOYSA-L | |
| hafnium(4+) bis(cyclopenta-2,4-dien-1-ide) dichloride | |
| 131674741 | |
| ≥98.0% (T) | |
| Hafnocene Dichloride |
| 233°C | |
| MFCD00001438,MFCD00001438 | |
| bis cyclopentadienyl hafniumdichloride | |
| [Cl-].[Cl-].[Hf+4].[CH-]1C=CC=C1.[CH-]1C=CC=C1 | |
| 379.58 | |
| 379.58 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : MG4815000
TSCA : Yes