missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hafnocene Dichloride 98.0+%, TCI America™
Supplier: TCI America H091425G
Specifications
| Hafnocene Dichloride | |
| 233°C | |
| MFCD00001438,MFCD00001438 | |
| bis cyclopentadienyl hafniumdichloride | |
| [Cl-].[Cl-].[Hf+4].[CH-]1C=CC=C1.[CH-]1C=CC=C1 | |
| 379.58 | |
| 379.58 | |
| Crystalline Powder |
| 12116-66-4 | |
| C10H10Cl2Hf | |
| 25 g | |
| CSEGCHWAMVIXSA-UHFFFAOYSA-L | |
| hafnium(4+) bis(cyclopenta-2,4-dien-1-ide) dichloride | |
| 131674741 | |
| ≥98.0% (T) |
Chemical Identifiers
| 12116-66-4 | |
| 379.58 | |
| CSEGCHWAMVIXSA-UHFFFAOYSA-L | |
| 131674741 | |
| [Cl-].[Cl-].[Hf+4].[CH-]1C=CC=C1.[CH-]1C=CC=C1 |
| C10H10Cl2Hf | |
| MFCD00001438,MFCD00001438 | |
| bis cyclopentadienyl hafniumdichloride | |
| hafnium(4+) bis(cyclopenta-2,4-dien-1-ide) dichloride |
Safety and Handling
RTECSNumber : MG4815000
TSCA : Yes