missing translation for 'onlineSavingsMsg'
Learn More

Hafnium standard solution, for AAS, 1 mg/ml Hf in 5% HF

Catalog No. AC196011000
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC196011000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC196011000 Supplier Thermo Scientific Chemicals Supplier No. 196011000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-58-6 | Hf | 178.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy

Specifications

Chemical Name or Material Hafnium standard solution
Packaging Plastic bottle
Name Note 1 mg/ml Hf in 5% HF
CAS 7440-58-6, 7664-39-3, 7732-18-5
Physical Form Solution
Molecular Formula Hf
Concentration 990 to 1010 μg/mL of Hf(4+) in 5% HF
MDL Number MFCD00011032
Color Yellow
Solubility Soluble in water
InChI Key VBJZVLUMGGDVMO-UHFFFAOYSA-N
SMILES [Hf]
IUPAC Name hafnium
Molecular Weight (g/mol) 178.49
Quantity 100 mL
PubChem CID 23986
ChEBI CHEBI:33343
Formula Weight 178.49
Grade AAS
Show More Show Less

RUO – Research Use Only