missing translation for 'onlineSavingsMsg'
Learn More

Hafnium(IV) Trifluoromethanesulfonate 90.0+%, TCI America™

Catalog No. T17081G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17081G 1 g
T17085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17081G Supplier TCI America Supplier No. T17081G
Only null left

Chemical Identifiers

CAS 161337-67-3
Molecular Formula C4H2F12HfO13S4
Molecular Weight (g/mol) 792.758
MDL Number MFCD01321255
InChI Key XQJOFTWVKHJLHA-UHFFFAOYSA-J
Synonym hafnium iv trifluoromethanesulfonate hydrate, hafnium tetrakis trifluoromethanesulfonate , hydrate
PubChem CID 71433309
IUPAC Name hafnium(4+);trifluoromethanesulfonate;hydrate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].O.[Hf+4]

Specifications

Color White-Yellow
Quantity 1 g
UN Number 1759
Formula Weight 774.74
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hafnium(IV) Trifluoromethanesulfonate