missing translation for 'onlineSavingsMsg'
Learn More

Hafnium(IV) sulfate, 99.9% (metals basis excluding Zr), Zr <1%

Catalog No. p-7059224
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4424406 5 g
AA4424414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4424406 Supplier Thermo Scientific Chemicals Supplier No. 04424406

CAS: 15823-43-5 | HfO4S | 274.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15823-43-5
Molecular Formula HfO4S
Molecular Weight (g/mol) 274.55
MDL Number MFCD00168081
InChI Key KOELDGKLEFTHGN-UHFFFAOYSA-L
Synonym hafnium sulfate, acmc-20aksa, sulfuric acid, hafnium 4+ salt 2:1, hafnium 4+ disulfate, hafnium iv sulfate tetrahydrate, sulfuric acid,hafnium 4+ salt 2:1, hafnium iv sulfate trace metals basis purity excludes zirconium
PubChem CID 13094115
IUPAC Name hafnium(4+);disulfate
SMILES [Hf].[O-]S([O-])(=O)=O

Specifications

Physical Form Powder
Quantity 5 g
Formula Weight 370.62 (Anhydrous)
Percent Purity 99.9%
Assay (metals basis excluding Zr), Zr <1%
Chemical Name or Material Hafnium (IV) sulfate
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only