Learn More
Hafnium(IV) oxide, 99.99% (metals basis excluding Zr), Zr <100ppm, Thermo Scientific Chemicals
$268.33 - $974.45
Chemical Identifiers
| CAS | 12055-23-1 |
|---|---|
| Molecular Formula | HfO2 |
| Molecular Weight (g/mol) | 210.49 |
| MDL Number | MFCD00003565 |
| InChI Key | WIHZLLGSGQNAGK-UHFFFAOYSA-N |
| IUPAC Name | hafnium(4+) dioxidandiide |
| SMILES | [O--].[O--].[Hf+4] |
Description
Hafnium(IV) oxide is used as Intermediates, Paint additives and coating additives, metal Products. And it is also used in optical coatings, as a refractory material in the insulation of such devices as thermocouples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12055-23-1 | |
| 210.49 | |
| WIHZLLGSGQNAGK-UHFFFAOYSA-N | |
| [O--].[O--].[Hf+4] |
| HfO2 | |
| MFCD00003565 | |
| hafnium(4+) dioxidandiide |
Specifications
| 2774°C | |
| Powder | |
| HfO2 | |
| 14,4588 | |
| WIHZLLGSGQNAGK-UHFFFAOYSA-N | |
| hafnium(4+) dioxidandiide | |
| 210.49 | |
| Odorless | |
| Hafnium(IV) oxide |
| 12055-23-1 | |
| 1 g | |
| MFCD00003565 | |
| Insoluble in water. | |
| [O--].[O--].[Hf+4] | |
| 210.49 | |
| 99.99% | |
| 9.68 g/mL | |
| (metals basis excluding Zr), Zr <100ppm |
Safety and Handling
EINECSNumber : 235-013-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only