missing translation for 'onlineSavingsMsg'
Learn More

Hafnium(IV) oxide, 99.9% (metals basis excluding Zr), Zr <0.5%, Thermo Scientific Chemicals

$70.26 - $587.85

Chemical Identifiers

CAS 12055-23-1
Molecular Formula HfO2
Molecular Weight (g/mol) 210.49
MDL Number MFCD00003565
InChI Key WIHZLLGSGQNAGK-UHFFFAOYSA-N
IUPAC Name hafnium(4+) dioxidandiide
SMILES [O--].[O--].[Hf+4]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3620404
View Documents
Thermo Scientific Chemicals
3620404
2 g
Each for $70.26
Only null left
 
AA3620409
View Documents
Thermo Scientific Chemicals
03620409
10 g
Each for $215.37
Only null left
 
AA3620418
View Documents
Thermo Scientific Chemicals
03620418
50 g
Each for $587.85
Only null left
 
Description

Description

Hafnium(IV) oxide is an intermediate. Hafnia is used in optical coatings, and as a high-κ dielectric in DRAM capacitors and in advanced metal-oxide-semiconductor devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12055-23-1
210.49
WIHZLLGSGQNAGK-UHFFFAOYSA-N
[O--].[O--].[Hf+4]
HfO2
MFCD00003565
hafnium(4+) dioxidandiide

Specifications

2774°C
Sintered Lump
HfO2
14,4588
WIHZLLGSGQNAGK-UHFFFAOYSA-N
hafnium(4+) dioxidandiide
210.49
Odorless
Hafnium(IV) oxide
12055-23-1
2 g
MFCD00003565
Insoluble in water.
[O--].[O--].[Hf+4]
210.49
99.9%
9.68 g/mL
(metals basis excluding Zr), Zr <0.5%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 235-013-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only