missing translation for 'onlineSavingsMsg'
Learn More

Hafnium(IV) oxide, 99.9% (metals basis excluding Zr), Zr <0.5%, Thermo Scientific Chemicals

Catalog No. AA3620404
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3620404 2 g
AA3620409 10 g
AA3620418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3620404 Supplier Thermo Scientific Chemicals Supplier No. 3620404

Hafnium(IV) oxide is an intermediate. Hafnia is used in optical coatings, and as a high-κ dielectric in DRAM capacitors and in advanced metal-oxide-semiconductor devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12055-23-1
Molecular Formula HfO2
Molecular Weight (g/mol) 210.49
MDL Number MFCD00003565
InChI Key WIHZLLGSGQNAGK-UHFFFAOYSA-N
IUPAC Name hafnium(4+) dioxidandiide
SMILES [O--].[O--].[Hf+4]

Specifications

Melting Point 2774°C
Physical Form Sintered Lump
Quantity 2 g
Merck Index 14,4588
Solubility Information Insoluble in water.
Formula Weight 210.49
Percent Purity 99.9%
Odor Odorless
Density 9.68 g/mL
Assay (metals basis excluding Zr), Zr <0.5%
Chemical Name or Material Hafnium(IV) oxide
Show More Show Less
EINECSNumber 235-013-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only