missing translation for 'onlineSavingsMsg'
Learn More

Hafnium boride, 99.5% (metals basis excluding Zr), Zr <2%, Thermo Scientific Chemicals

$570.19 - $570.19

Chemical Identifiers

CAS 12007-23-7
Molecular Formula B2Hf
MDL Number MFCD00016125
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4366418
View Documents
Thermo Scientific Chemicals
04366418
50 g
Each for $570.19
Only null left
 
Description

Description

Hafnium boride is used in wear resistant coatings. It is also employed as a nuclear reactor control rod. Furthermore, it is used in hypervelocity re-entry vehicles like ICBM heat shields or aerodynamic leading-edges. It is useful in the preparation of high temperature composites in conjunction with silicon carbide (SiC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12007-23-7
B2Hf
MFCD00016125

Specifications

3250°C
Powder
B2Hf
MFCD00016125
200.11
Odorless
Hafnium boride
12007-23-7
50 g
HfB2
Insoluble in water.
99.5%
10.5 g/mL
(metals basis excluding Zr), Zr <2%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 234-500-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only