accessibility menu, dialog, popup

UserName

CAS RN 12007-23-7

BBHf

CAS RN 12007-23-7

IUPAC Name: diboron hafnium
Synonyms: hafnium diboride
Molecular Weight (g/mol): 200.11
Molecular Formula: B2Hf
InChi Key: LRTTZMZPZHBOPO-UHFFFAOYSA-N
SMILES: [B].[B].[Hf]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results