Learn More
H-B Instrument™ Durac™ Salt Brine Plain Form Hydrometers
Choose from two plain form models with ranges from 0 to 100% or 0 to 26.5% to measure sodium chloride saturation of salt brines
Description
- Lead-free ballast and glass
- Individually serialized
- Temperature standardization: 60°F (15°C)
Order Info
Should you require an individual Hydrometer Calibration Report, factory calibrated hydrometers or custom calibration services are available.
Certifications
Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team.
Chemical Identifiers
| Traceable to NIST | |
| Salt Brine hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures. | |
| Used in various industries including food, fishing, and transportation. |
Specifications
| Hydrometer (% by Saturation) | |
| 1% | |
| 280 mm | |
| Used in various industries including food, fishing, and transportation. |
| ±1% | |
| Traceable to NIST | |
| Salt Brine hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures. | |
| 0 to 100% |