missing translation for 'onlineSavingsMsg'
Learn More

GW 9662, Thermo Scientific Chemicals

Catalog No. p-7043123
Change view
Click to view available options
Quantity:
10 mg
Catalog No. Quantity
AAJ65634MA 10 mg
1 options

Catalog No. AAJ65634MA

Supplier: Thermo Scientific Chemicals J65634MA

Only null left

An irreversible PPAR→ antagonist

An irreversible PPAR- antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22978-25-2
Molecular Formula C13H9ClN2O3
Molecular Weight (g/mol) 276.676
MDL Number MFCD01215270
InChI Key DNTSIBUQMRRYIU-UHFFFAOYSA-N
Synonym 2-chloro-5-nitrobenzanilide, 2-chloro-5-nitro-n-phenyl-benzamide, 2-chloro-5-nitro-n-4-phenylbenzamide, 2-chloro-5-nitrophenyl-n-benzamide, tocris-1508, spectrum5_001989, lopac-m-6191, d00acx, d05thb, dsstox_cid_20723
PubChem CID 644213
ChEBI CHEBI:79993
IUPAC Name 2-chloro-5-nitro-N-phenylbenzamide
SMILES C1=CC=C(C=C1)NC(=O)C2=C(C=CC(=C2)[N+](=O)[O-])Cl

Specifications

Color White
Quantity 10 mg
Formula Weight 276.68
Physical Form Powder
Chemical Name or Material GW 9662
Hazard Category H317-H319
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only