missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals GW-405833

Catalog No. AAJ67425MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67425MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67425MA Supplier Thermo Scientific Chemicals Supplier No. J67425MA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 180002-83-9 | C23H24Cl2N2O3, C23H24Cl2N2O3 | 447.36 g/mol

It is a potent and selective CB2 receptor partial agonist (EC50 = 0.65 nM; maximum inhibition = 44.6%). Binds with high affinity to both human and rat CB2 receptors and displays ∼ 1200-fold selectivity over CB1 (Ki values are 3.92 and 4772 nM for human recombinant CB2 and CB1 receptors respectively). Produces potent antihyperalgesic effects in several rodent models of pain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 180002-83-9
Molecular Formula C23H24Cl2N2O3
Synonym L-768242; 1-(2,3-Dichlorobenzoyl)-5-methoxy-2-methyl-3-[2-(4-morpholinyl)ethyl]indole

Specifications

Quantity 10 mg
Description Off-white
Content And Storage -30°C to -10°C
Formula Weight 447.36
Melting Point 47°C to 49°C
Solubility Soluble in DMSO at 10-15mg/ml. Also soluble in ethanol at ∽5mg/ml.
Product Type GW-405833
Form Solid

RUO – Research Use Only