accessibility menu, dialog, popup

UserName

CAS RN 180002-83-9

IUPAC Name: 1-(2,3-dichlorobenzoyl)-5-methoxy-2-methyl-3-[2-(morpholin-4-yl)ethyl]-1H-indole
Synonyms: 1-(2,3-dichlorobenzoyl)-5-methoxy-2-methyl-3-[2-(morpholin-4-yl)ethyl]indole
Molecular Weight (g/mol): 447.36
Molecular Formula: C23H24Cl2N2O3
InChi Key: FSFZRNZSZYDVLI-UHFFFAOYSA-N
SMILES: COC1=CC=C2N(C(=O)C3=C(Cl)C(Cl)=CC=C3)C(C)=C(CCN3CCOCC3)C2=C1
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results