missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GW 2580 98.0+%, TCI America™
$410.10 - $1217.22
Chemical Identifiers
| CAS | 870483-87-7 |
|---|---|
| Molecular Formula | C20H22N4O3 |
| Molecular Weight (g/mol) | 366.42 |
| MDL Number | MFCD12024701 |
| InChI Key | MYQAUKPBNJWPIE-UHFFFAOYSA-N |
| Synonym | 5-[3-Methoxy-4-[(4-methoxybenzyl)oxy]benzyl]pyrimidine-2,4-diamine |
| PubChem CID | 11617559 |
| IUPAC Name | 5-({3-methoxy-4-[(4-methoxyphenyl)methoxy]phenyl}methyl)pyrimidine-2,4-diamine |
| SMILES | COC1=CC=C(COC2=C(OC)C=C(CC3=CN=C(N)N=C3N)C=C2)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
G050450MG
|
TCI America
G050450MG |
50 mg |
Each for $410.10
|
|
|||||
|
G0504250MG
|
TCI America
G0504250MG |
250 mg |
Each for $1,217.22
|
|
|||||
Chemical Identifiers
| 870483-87-7 | |
| 366.42 | |
| MYQAUKPBNJWPIE-UHFFFAOYSA-N | |
| 11617559 | |
| COC1=CC=C(COC2=C(OC)C=C(CC3=CN=C(N)N=C3N)C=C2)C=C1 |
| C20H22N4O3 | |
| MFCD12024701 | |
| 5-[3-Methoxy-4-[(4-methoxybenzyl)oxy]benzyl]pyrimidine-2,4-diamine | |
| 5-({3-methoxy-4-[(4-methoxyphenyl)methoxy]phenyl}methyl)pyrimidine-2,4-diamine |
Specifications
| 870483-87-7 | |
| MFCD12024701 | |
| 5-[3-Methoxy-4-[(4-methoxybenzyl)oxy]benzyl]pyrimidine-2,4-diamine | |
| COC1=CC=C(COC2=C(OC)C=C(CC3=CN=C(N)N=C3N)C=C2)C=C1 | |
| 366.42 | |
| 366.42 | |
| Crystalline Powder |
| C20H22N4O3 | |
| 50 mg | |
| MYQAUKPBNJWPIE-UHFFFAOYSA-N | |
| 5-({3-methoxy-4-[(4-methoxyphenyl)methoxy]phenyl}methyl)pyrimidine-2,4-diamine | |
| 11617559 | |
| ≥98.0% (HPLC) | |
| GW 2580 |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)