Learn More
GVS Nitrocellulose Hybridization and Transfer Membranes: Circles
Require vacuum baking to fix DNA
Supplier: GVS 1215473
Description
Nitrocellulose Unsupported Membrane (EP)
- Highest sensitivity for protein blotting and immunoblotting, or other procedures requiring optimum resolution
- Easily blocked
- Wets out naturally and does not require methanol like PVDF membranes
- High sensitivity but with added strength of a polyester matrix
- Surfactant-free membranes have no additives to interfere with binding capacity
Specifications
| Transfer Membrane | |
| Disc | |
| 0.45 μm |
| 85 mm | |
| NitroBind |