missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanylurea Phosphate 98.0+%, TCI America™
$69.31 - $410.07
Chemical Identifiers
| CAS | 36897-89-9 |
|---|---|
| Molecular Formula | C2H9N4O5P |
| Molecular Weight (g/mol) | 200.09 |
| MDL Number | MFCD00065097 |
| InChI Key | ZHURYQINAXWNAH-UHFFFAOYSA-N |
| Synonym | Dicyandiamidine Phosphate |
| IUPAC Name | (diaminomethylidene)urea; phosphoric acid |
| SMILES | OP(O)(O)=O.NC(N)=NC(N)=O |
Chemical Identifiers
| 36897-89-9 | |
| 200.09 | |
| ZHURYQINAXWNAH-UHFFFAOYSA-N | |
| (diaminomethylidene)urea; phosphoric acid |
| C2H9N4O5P | |
| MFCD00065097 | |
| Dicyandiamidine Phosphate | |
| OP(O)(O)=O.NC(N)=NC(N)=O |
Specifications
| 36897-89-9 | |
| White | |
| MFCD00065097 | |
| Dicyandiamidine Phosphate | |
| OP(O)(O)=O.NC(N)=NC(N)=O | |
| 200.09 | |
| ≥98.0% (T) | |
| Guanylurea Phosphate |
| 184°C | |
| C2H9N4O5P | |
| 25 g | |
| ZHURYQINAXWNAH-UHFFFAOYSA-N | |
| (diaminomethylidene)urea; phosphoric acid | |
| 200.09 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1778
TSCA : No