accessibility menu, dialog, popup

UserName

NH2NH2NNH2OOHPOHOHO

CAS RN 36897-89-9

IUPAC Name: (diaminomethylidene)urea; phosphoric acid
Synonyms: diaminomethylideneurea; phosphoric acid
Molecular Weight (g/mol): 200.09
Molecular Formula: C2H9N4O5P
InChi Key: ZHURYQINAXWNAH-UHFFFAOYSA-N
SMILES: OP(O)(O)=O.NC(N)=NC(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results