missing translation for 'onlineSavingsMsg'
Learn More

Guanylurea Phosphate 98.0+%, TCI America™

Catalog No. D0432500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0432500G 500 g
D043225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0432500G Supplier TCI America Supplier No. D0432500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 36897-89-9
Molecular Formula C2H9N4O5P
Molecular Weight (g/mol) 200.09
MDL Number MFCD00065097
InChI Key ZHURYQINAXWNAH-UHFFFAOYSA-N
Synonym Dicyandiamidine Phosphate
IUPAC Name (diaminomethylidene)urea; phosphoric acid
SMILES OP(O)(O)=O.NC(N)=NC(N)=O

Specifications

Melting Point 184°C
Color White
Quantity 500 g
Formula Weight 200.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Guanylurea Phosphate
EINECSNumber (2)-1778
TSCA No