missing translation for 'onlineSavingsMsg'
Learn More

Guanosine 98.0+%, TCI America™

Catalog No. p-7130176
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Catalog No. Quantity
G01715G 5 g
G017125G 25 g
G0171100G 100 g
3 options

Catalog No. G01715G

Supplier: TCI America G01715G

Only null left

Chemical Identifiers

CAS 118-00-3
Molecular Formula C10H13N5O5
Molecular Weight (g/mol) 283.24
MDL Number MFCD00010182
InChI Key NYHBQMYGNKIUIF-UUOKFMHZSA-N
Synonym guanosine, guanine riboside, vernine, guanozin, guanosin, inosine, 2-amino, usaf cb-11, vernine van, l-guanosine, 9-beta-d-ribofuranosylguanine
PubChem CID 6802
ChEBI CHEBI:16750
IUPAC Name 2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
SMILES NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1

Specifications

Color White
Quantity 5 g
Formula Weight 283.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Guanosine
EINECSNumber (8)-0279
RTECSNumber MF8750000
TSCA Yes