accessibility menu, dialog, popup

UserName

N(R)NNHN(R)O(S)(R)OH2NOHHOHO

CAS RN 118-00-3

IUPAC Name: 2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Synonyms: guanosine|guanine riboside|Guo
Molecular Weight (g/mol): 283.24
Molecular Formula: C10H13N5O5
InChi Key: NYHBQMYGNKIUIF-UUOKFMHZSA-N
SMILES: NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1
Percent Purity 
  • (4)
  • (3)
  • (3)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (7)
  • (4)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)