missing translation for 'onlineSavingsMsg'
Learn More

Guanosine, 98%

CAS: 118-00-3 | C10H13N5O5 | 283.24 g/mol

Supplier:  Thermo Scientific Chemicals A1132818

 View more versions of this product

Catalog No. AAA1132818


Only null left
Explore available promotions

Description

Description

Guanosine is used as a constituent of nucleic acids. It is used in metallic paints, simulated pearls,plastics,cosmetics industry etc,. It has been also used in pharmacokinetics as a prodrug. Guanosine is used in cell culture applications as a precursor of GMP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Guanosine is used as a constituent of nucleic acids. It is used in metallic paints, simulated pearls,plastics,cosmetics industry etc,. It has been also used in pharmacokinetics as a prodrug. Guanosine is used in cell culture applications as a precursor of GMP.

Solubility
Soluble in Formic Acid:Water (1:1): 50 mg/mL. Insoluble in methanol, diethyl ether.

Notes
Store away from extreme heat. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Guanosine
∼250°C (decomposition)
MFCD00010182
625911
guanosine, guanine riboside, vernine, guanozin, guanosin, inosine, 2-amino, usaf cb-11, vernine van, l-guanosine, 9-beta-d-ribofuranosylguanine
NYHBQMYGNKIUIF-UUOKFMHZSA-N
NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1
283.24
CHEBI:16750
≥98%
118-00-3
C10H13N5O5
50 g
14,4566
Soluble in Formic Acid:Water (1:1): 50mg/mL. Insoluble in methanol,diethyl ether.
−72° (c=1.5 in 0.1N NaOH)
2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
6802
283.24

Chemical Identifiers

118-00-3
283.24
NYHBQMYGNKIUIF-UUOKFMHZSA-N
6802
NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1
C10H13N5O5
MFCD00010182
guanosine, guanine riboside, vernine, guanozin, guanosin, inosine, 2-amino, usaf cb-11, vernine van, l-guanosine, 9-beta-d-ribofuranosylguanine
CHEBI:16750
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 204-227-8

RTECSNumber : MF8750000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only