missing translation for 'onlineSavingsMsg'
Learn More

Guanine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015377
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015377 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015377 Supplier Merck Emd Millipore Supplier No. PHR1243500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Guanine is one of the building blocks of both RNA and DNA that plays a significant role in protein biosynthesis as well as the storage of genetic information. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 73-40-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C5H5N5 O
MDL Number MFCD00071533
Melting Point >300°C
Synonym 2-Amino-1,7-dihydro-6 H-purin-6-one; 2-Amino-6-hydroxypurine; 2-Aminohypoxanthine
Type Certified Reference Material
Quantity 500 mg
Formula Weight 151.13
Grade Certified Reference Material
Show More Show Less