missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Guanidine Thiocyanate, ≥99%, Calbiochem™,

$164.14 - $712.00

Chemical Identifiers

CAS 593-84-0
Molecular Formula CH5N3 · CHNS
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
368975100GM
View Documents Promotion Details
MilliporeSigma™
368975100GM
100 g Glass Bottle
Each for $164.14
Only null left
 
368975500GM
View Documents Promotion Details
MilliporeSigma™
368975500GM
500 g Glass Bottle
Each for $712.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

593-84-0
CH5N3 · CHNS

Specifications

White
593-84-0
CH5N3 · CHNS
≥99% (Titration)
Videos
SDS
Documents

Documents