Learn More
Thermo Scientific Chemicals Guanidine thiocyanate, 99%
Supplier: Thermo Scientific Chemicals B212500E
Description
Guanidine thiocyanate is a useful tool for RNA isolation and protein solubilization. It is involved in the quantification of mRNA from hepatocytes. It acts as a medium for blood smaple lystae in order to maintain the integrity of nucleic acid. It is a chaotropic agent, mainly involved in the extraction of DNA and RNA. It is utilized to deactivate the influenza virus.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGuanidine thiocyanate is a useful tool for RNA isolation and protein solubilization. It is involved in the quantification of mRNA from hepatocytes. It acts as a medium for blood smaple lystae in order to maintain the integrity of nucleic acid. It is a chaotropic agent, mainly involved in the extraction of DNA and RNA. It is utilized to deactivate the influenza virus.
Solubility
Soluble in water and ethanol
Notes
Light sensitive. Incompatible with acids and strong oxidizing agents.
Specifications
| Guanidine thiocyanate | |
| 2500 g | |
| 99% |
| Liquid | |
| Soluble in water and ethanol |
Safety and Handling
Recommended Storage : Ambient temperatures
RUO – Research Use Only