missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanidine Phosphate 95.0+%, TCI America™
$48.46 - $208.78
Chemical Identifiers
| CAS | 5423-23-4 |
|---|---|
| Molecular Formula | CH8N3O4P |
| Molecular Weight (g/mol) | 157.066 |
| MDL Number | MFCD00039131 |
| InChI Key | CEDDGDWODCGBFQ-UHFFFAOYSA-N |
| PubChem CID | 160593 |
| IUPAC Name | guanidine;phosphoric acid |
| SMILES | C(=N)(N)N.OP(=O)(O)O |
Chemical Identifiers
| 5423-23-4 | |
| 157.066 | |
| CEDDGDWODCGBFQ-UHFFFAOYSA-N | |
| guanidine;phosphoric acid |
| CH8N3O4P | |
| MFCD00039131 | |
| 160593 | |
| C(=N)(N)N.OP(=O)(O)O |
Specifications
| 5423-23-4 | |
| CH8N3O4P | |
| 25 g | |
| C(=N)(N)N.OP(=O)(O)O | |
| 157.066 | |
| 216.14 | |
| Crystalline Powder |
| White | |
| MFCD00039131 | |
| CEDDGDWODCGBFQ-UHFFFAOYSA-N | |
| guanidine;phosphoric acid | |
| 160593 | |
| ≥95.0% (T) | |
| Guanidine Phosphate |
Safety and Handling
TSCA : No