accessibility menu, dialog, popup

UserName

CAS RN 5423-23-4

NH2NH2HNNH2NH2HNOHPOHOHO

CAS RN 5423-23-4

IUPAC Name: bis(guanidine); phosphoric acid
Synonyms: bis(guanidine); phosphoric acid
Molecular Weight (g/mol): 216.14
Molecular Formula: C2H13N6O4P
InChi Key: SSHPJTCTVWVCLI-UHFFFAOYSA-N
SMILES: NC(N)=N.NC(N)=N.OP(O)(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results