missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanidine Phosphate 95.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 500 g |
| Formula Weight | 216.14 |
| Percent Purity | ≥95.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Guanidine Phosphate |
Safety and Handling
| TSCA | No |