missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrochloride 99.0+%, TCI America™

Catalog No. p-7130186
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G019725G 25 g
G0197100G 100 g
G0197500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G019725G Supplier TCI America Supplier No. G019725G
Only null left

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
MDL Number MFCD00013026
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Melting Point 187°C
Color White
pH 5.56 (50g/L H2O soln. 25°C)
Quantity 25 g
Formula Weight 95.53
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Guanidine Hydrochloride [for Protein Research]
RTECSNumber MF4300000
TSCA Yes