missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanidine Hydrochloride, (Crystalline, solution may be cloudy and yellow/Practical Grade), MP Biomedicals
$151.21 - $567.19
Chemical Identifiers
| CAS | 50-01-1 |
|---|---|
| Molecular Formula | CH6ClN3 |
| Molecular Weight (g/mol) | 95.53 |
| InChI Key | PJJJBBJSCAKJQF-UHFFFAOYSA-N |
| Synonym | guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride |
| PubChem CID | 5742 |
| ChEBI | CHEBI:32735 |
| IUPAC Name | guanidine;hydrochloride |
| SMILES | C(=N)(N)N.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10190501
|
MP Biomedicals Inc
0210190501 |
1 kg |
Each for $151.21
|
|
|||||
|
ICN10190505
|
MP Biomedicals Inc
0210190505 |
5 kg |
Each for $567.19
|
|
|||||
Chemical Identifiers
| 50-01-1 | |
| 95.53 | |
| guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
| CHEBI:32735 | |
| C(=N)(N)N.Cl |
| CH6ClN3 | |
| PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
| 5742 | |
| guanidine;hydrochloride |
Specifications
| 50-01-1 | |
| CH6ClN3 | |
| guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
| C(=N)(N)N.Cl | |
| 95.53 | |
| CHEBI:32735 | |
| Practical | |
| Guanidine Hydrochloride |
| Yellow | |
| 1 kg | |
| PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
| guanidine;hydrochloride | |
| 5742 | |
| 95.5 | |
| Crystalline, Solution May Be Cloudy |
Safety and Handling
Recommended Storage : Room Temperature