missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrochloride, (Crystalline, solution may be cloudy and yellow/Practical Grade), MP Biomedicals

Catalog No. ICN10190501
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10190501 1 kg
ICN10190505 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10190501 Supplier MP Biomedicals Inc Supplier No. 0210190501

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Color Yellow
Quantity 1 kg
Formula Weight 95.5
Grade Practical
Physical Form Crystalline, Solution May Be Cloudy
Chemical Name or Material Guanidine Hydrochloride
Recommended Storage Room Temperature